(3-Fluorophenyl)acetonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

F0256-25G F0256-5G
TCIAF0256-25GEA 101 EUR
TCIAF0256-25G TCIAF0256-5G
(3-Fluorophenyl)acetonitrile ≥98.0% (by GC)
(3-Fluorophenyl)acetonitrile
Formule: C₈H₆FN
Kookpunt: 230°C
Dichtheid: 1,14058 g/cm³ (15 °C)
MDL Nummer: MFCD00001907
CAS nummer: 501-00-8
EINECS: 207-918-2
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64089203d67fb3dd579d37d6a261b2b6d427e759&vid=64089203d67fb3dd579d37d6a261b2b6d427e759&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR