4'-Fluoropropiophenone ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: p-Fluoropropiophenone, 1-(4-Fluorophenyl)propan-1-one

F0522-25G F0522-5G
TCIAF0522-25GEA 48.4 EUR
TCIAF0522-25G TCIAF0522-5G
4'-Fluoropropiophenone ≥97.0% (by GC)
4'-Fluoropropiophenone
Formule: C₉H₉FO
Kookpunt: 210°C
Dichtheid: 1,09 g/cm³ (20 °C)
Vlampunt: 76°C
MDL Nummer: MFCD00000356
CAS nummer: 456-03-1
EINECS: 207-255-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=465c16fa2cbf6a1841c181202acf741fcc178ef3&vid=465c16fa2cbf6a1841c181202acf741fcc178ef3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR