4'-Iodoacetophenone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

I0305-25G I0305-5G
TCIAI0305-25GEA 84.7 EUR
TCIAI0305-25G TCIAI0305-5G
4'-Iodoacetophenone ≥98.0% (by GC)
4'-Iodoacetophenone
Formule: C₈H₇IO
Kookpunt: 145 - 150 °C (20 -rr)
Smeltpunt: 80…84 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00045320
CAS nummer: 13329-40-3
EINECS: 236-372-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR