4-Iodobenzyl alcohol ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
I0810-5G I0810-25G
TCIAI0810-5GEA 46.2 EUR
TCIAI0810-5G TCIAI0810-25G
4-Iodobenzyl alcohol ≥99.0% (by GC)
4-Iodobenzyl alcohol
Formule: C₇H₇IO
Kookpunt: 136°C
MDL Nummer: MFCD01732720
CAS nummer: 18282-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25e492511591c0e1e690201ec3db1d8b29121c22&vid=25e492511591c0e1e690201ec3db1d8b29121c22&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 71.0 to 75.0 deg-C