4-Hydroxybenzyl alcohol ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (4-Hydroxyphenyl)methanol, p-Hydroxybenzyl Alcohol, 4-(Hydroxymethyl)phenol

H0224-25G H0224-100G
TCIAH0224-25GEA 27.5 EUR
TCIAH0224-25G TCIAH0224-100G
4-Hydroxybenzyl alcohol ≥97.0% (by GC)
4-Hydroxybenzyl alcohol
Formule: C₇H₈O₂
MW: 124.14 g/mol
Kookpunt: 252
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004658
CAS nummer: 623-05-2
EINECS: 210-768-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bbf89cf74dc506d961fe7881f0c32588ee86644&vid=2bbf89cf74dc506d961fe7881f0c32588ee86644&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR