1-Propyl-4-ethynylbenzene ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Ethynyl-4-propylbenzene, (4-Propylphenyl)acetylene, 4-n-Propylphenylacetylene, (4-Propylphenyl)ethyne
E0750-25G E0750-5G
TCIAE0750-25GEA 85.8 EUR
TCIAE0750-25G TCIAE0750-5G
1-Propyl-4-ethynylbenzene ≥96.0% (by GC)
1-Propyl-4-ethynylbenzene
Formule: C₁₁H₁₂
MDL Nummer: MFCD00173886
CAS nummer: 62452-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c368f23882a3f41fed61ce2d213d702e988b02d0&vid=c368f23882a3f41fed61ce2d213d702e988b02d0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 96.0 %