3-Benzyloxybenzyl alcohol ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B3857-25G B3857-5G
TCIAB3857-25GEA 259 EUR
TCIAB3857-25G TCIAB3857-5G
3-Benzyloxybenzyl alcohol ≥97.0% (by GC)
3-Benzyloxybenzyl alcohol
Formule: C₁₄H₁₄O₂
Kookpunt: 160°C
Smeltpunt: 48 °C
MDL Nummer: MFCD00004642
CAS nummer: 1700-30-7
EINECS: 216-931-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc13c8170eb6f8d37fffc20b2c8771bb6a1166c9&vid=bc13c8170eb6f8d37fffc20b2c8771bb6a1166c9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 47.0 to 51.0 deg-C