3-(Trifluoromethyl)benzyl alcohol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
T1342-10G
T1342-25G
TCIAT1342-10GEA
53.9
EUR
TCIAT1342-10G
TCIAT1342-25G
3-(Trifluoromethyl)benzyl alcohol ≥98.0% (by GC)
3-(Trifluoromethyl)benzyl alcohol
Formule:
C₈H₇F₃O Kookpunt: 68°C Dichtheid: 1,2949 g/cm³ (21 °C) Vlampunt: 84°C |
MDL Nummer:
MFCD00004645 CAS nummer: 349-75-7 EINECS: 206-489-9 |
|
Specification Test Results
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...