4,4'-Difluorobiphenyl
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D2181-1G
D2181-5G
TCIAD2181-1GEA
33
EUR
TCIAD2181-1G
TCIAD2181-5G
4,4'-Difluorobiphenyl
4,4'-Difluorobiphenyl
Formule:
C₁₂H₈F₂ Kookpunt: 255°C Smeltpunt: 95 °C Dichtheid: 1,361 g/cm³ (20 °C) |
MDL Nummer:
MFCD00000349 CAS nummer: 398-23-2 EINECS: 206-910-6 UN: 3152 ADR: 9,II Merck Index: 14,03149 |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 91.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...