Cyclopentanecarbonyl chloride ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Cyclopentanecarboxylic chloride

C2391-25G C2391-5G
TCIAC2391-25GEA 200 EUR
TCIAC2391-25G TCIAC2391-5G
Cyclopentanecarbonyl chloride ≥98.0% (by GC, titration analysis)
Cyclopentanecarbonyl chloride
Formule: C₆H₉ClO
MW: 132.59 g/mol
Kookpunt: 161°C
Vlampunt: 60°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001370
CAS nummer: 4524-93-0
EINECS: 224-856-1
UN: 2920
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29e01436f112b4158e09a3218c025d1b2ea54bd7&vid=29e01436f112b4158e09a3218c025d1b2ea54bd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR