Octaphenylcyclotetrasiloxane ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
O0231-25G
O0231-5G
TCIAO0231-25GEA
48.4
EUR
TCIAO0231-25G
TCIAO0231-5G
Octaphenylcyclotetrasiloxane ≥98.0% (by HPLC)
Octaphenylcyclotetrasiloxane
Formule:
C₄₈H₄₀O₄Si₄ Kookpunt: 330 - 340 °C (1013 hPa) Smeltpunt: 198…200 °C Dichtheid: 1,089 g/cm³ (20 °C) |
MDL Nummer:
MFCD00003268 CAS nummer: 546-56-5 EINECS: 208-904-9 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 198.0 to 205.0 deg-C |
Solubility in Toluene | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...