Tris(2-methylphenyl)phosphine ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Tris(o-tolyl)phosphine, Tri-o-tolylphosphine
T1024-25G T1024-5G
TCIAT1024-25GEA 120 EUR
TCIAT1024-25G TCIAT1024-5G
Tris(2-methylphenyl)phosphine ≥97.0% (by GC)
Tris(2-methylphenyl)phosphine
Formule: C₂₁H₂₁P
Kookpunt: 187 - 192 °C (0,05 -rr)
MDL Nummer: MFCD00008514
CAS nummer: 6163-58-2
EINECS: 228-193-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e52ae7931133d29028afe6a6ef234ff0c9d086f&vid=1e52ae7931133d29028afe6a6ef234ff0c9d086f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 124.0 to 129.0 deg-C