1-Naphthoic acid ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
N0024-25G N0024-500G
TCIAN0024-25GEA 47.3 EUR
TCIAN0024-25G TCIAN0024-500G
1-Naphthoic acid ≥98.0% (by titrimetric analysis)
1-Naphthoic acid
Formule: C₁₁H₈O₂
Kookpunt: 300 °C (1013 hPa)
Smeltpunt: 162 °C
Dichtheid: 1 g/cm³ (20 °C)
MDL Nummer: MFCD00004007
CAS nummer: 86-55-5
EINECS: 201-681-9
Merck Index: 14,06381
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdcc312e2fbc4bf0ee7c07c6d135bec38855f59a&vid=fdcc312e2fbc4bf0ee7c07c6d135bec38855f59a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 164.0 deg-C