Acenaphthenequinone ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A0004-25G
A0004-100G
TCIAA0004-25GEA
165
EUR
TCIAA0004-25G
TCIAA0004-100G
Acenaphthenequinone ≥98.0% (by GC)
Acenaphthenequinone
Formule:
C₁₂H₆O₂ Kookpunt: 351 °C (1013 hPa) Smeltpunt: 252…255 °C Dichtheid: 1,48 g/cm³ (20 °C) |
MDL Nummer:
MFCD00003805 CAS nummer: 82-86-0 EINECS: 201-441-3 |
|
Specification Test Results
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in hot DMF | within almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...