2-Chloroisonicotinic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Chloro-4-pyridinecarboxylic acid, 2-Chloropyridine-4-carboxylic acid

C1368-25G C1368-5G
TCIAC1368-25GEA 135 EUR
TCIAC1368-25G TCIAC1368-5G
2-Chloroisonicotinic acid ≥98.0% (by HPLC, titration analysis)
2-Chloroisonicotinic acid
Formule: C₆H₄ClNO₂
MDL Nummer: MFCD00191402
CAS nummer: 6313-54-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d45e5de746ec19086fde17538f9f26ff9048ba42&vid=d45e5de746ec19086fde17538f9f26ff9048ba42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in 1mol/L NaOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR