Ethyl isonicotinate ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
I0137-25ML
I0137-500ML
TCIAI0137-25MLEA
37.4
EUR
TCIAI0137-25ML
TCIAI0137-500ML
Ethyl isonicotinate ≥98.0% (by GC, titration analysis)
Ethyl isonicotinate
Formule:
C₈H₉NO₂ Kookpunt: 220 °C (1013 hPa) Dichtheid: 1,1 g/cm³ (20 °C) Vlampunt: 87°C |
MDL Nummer:
MFCD00006428 CAS nummer: 1570-45-2 EINECS: 216-379-2 Merck Index: 13,05206 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...