1,1,3,3-Tetramethyldisiloxane ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
T1437-25ML T1437-250ML
TCIAT1437-25MLEA 19.8 EUR
TCIAT1437-25ML TCIAT1437-250ML
1,1,3,3-Tetramethyldisiloxane ≥97.0% (by GC)
1,1,3,3-Tetramethyldisiloxane
Formule: C₄H₁₄OSi₂
MW: 134.33 g/mol
Kookpunt: 71°C
Dichtheid: 0.7555
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008256
CAS nummer: 3277-26-7
EINECS: 221-906-4
UN: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cc5a7b6f2b000a2a83a778934719c037801925e&vid=2cc5a7b6f2b000a2a83a778934719c037801925e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %