3-Methoxythiophene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M1093-1G M1093-5G
TCIAM1093-1GEA 17.6 EUR
TCIAM1093-1G TCIAM1093-5G
3-Methoxythiophene ≥98.0% (by GC)
3-Methoxythiophene
Formule: C₅H₆OS
Kookpunt: 82°C
Smeltpunt: –7 °C
Dichtheid: 1,385 g/cm³ (-173.15 °C)
Vlampunt: 49°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00043894
CAS nummer: 17573-92-1
EINECS: 479-350-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e45362873e44932249379554ffec1bd537154bce&vid=e45362873e44932249379554ffec1bd537154bce&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %