2-Chloro-3-methylthiophene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C1801-25G C1801-5G
TCIAC1801-25GEA 137 EUR
TCIAC1801-25G TCIAC1801-5G
2-Chloro-3-methylthiophene ≥98.0% (by GC)
2-Chloro-3-methylthiophene
Formule: C₅H₅ClS
MW: 132.61 g/mol
Kookpunt: 54°C
Dichtheid: 1.2281
MDL Nummer: MFCD00130083
CAS nummer: 14345-97-2
EINECS: 238-296-0
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca57f56b35c13cb84006c0feaadda7e969e7e97a&vid=ca57f56b35c13cb84006c0feaadda7e969e7e97a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %