2-Acetylthiophene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Thiophen-2-yl-ethanone, 1-(2-Thienyl)-1-ethanone, Methyl 2-thienyl ketone
A0724-25G A0724-500G A0724-100G
TCIAA0724-25GEA 18.7 EUR
TCIAA0724-25G TCIAA0724-500G TCIAA0724-100G
2-Acetylthiophene ≥98.0% (by GC)
2-Acetylthiophene
Formule: C₆H₆OS
Kookpunt: 214°C
Smeltpunt: 9 °C
Dichtheid: 1,1704 g/cm³ (20 °C)
Vlampunt: 91°C
MDL Nummer: MFCD00005442
CAS nummer: 88-15-3
EINECS: 201-804-6
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4895e631a434b1a78878c45fc2cf9d73e7c6148&vid=f4895e631a434b1a78878c45fc2cf9d73e7c6148&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %