Xanthan gum
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
X0048-25G
X0048-500G
X0048-100G
TCIAX0048-25GEA
17.6
EUR
TCIAX0048-25G
TCIAX0048-500G
TCIAX0048-100G
Xanthan gum
Xanthan gum
Dichtheid:
1,5 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00131256 CAS nummer: 11138-66-2 EINECS: 234-394-2 Merck Index: 14,10057 |
Specification Test Results
Appearance | White to Light red to Green powder to crystal |
Viscosity | 1,450 to 2,000 mPa-s (1 %, H2O, 20 deg-C) (add 1%KCl, calcd.on dried substance) |
Drying loss | max. 15.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...