Dodecamolybdophosphoric acid hydrate
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate
P1910-100G P1910-500G P1910-25G
TCIAP1910-100GEA 71.5 EUR
TCIAP1910-100G TCIAP1910-500G TCIAP1910-25G
Dodecamolybdophosphoric acid hydrate
Dodecamolybdophosphoric acid hydrate
Formule: H₃[P(Mo₃O₁₀)₄]·nH₂O
Smeltpunt: 78…90 °C
Dichtheid: 1,62 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149913
CAS nummer: 51429-74-4
EINECS: 234-713-5
UN: 3085
ADR: 5.1,III
Merck Index: 12,07498
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40d1d77133aaf6f819deea9b9604bb8b5b9bc667&vid=d8ac726ab77d7276f92eed773e2cc7ac20548bd7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Molybdenum oxide min. 72.0 %