(R)-(+)-1-Phenylethyl isocyanate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: (R)-(+)-α-Phenethyl isocyanate, (R)-(+)-1-Phenethyl isocyanate, (R)-(+)-1-phenylethylisocyanate, (R)-(+)-α-Methylbenzyl isocyanate

I0334-1G I0334-25G I0334-5G
TCIAI0334-1GEA 33 EUR
TCIAI0334-1G TCIAI0334-25G TCIAI0334-5G
(R)-(+)-1-Phenylethyl isocyanate ≥98.0% (by GC)
(R)-(+)-1-Phenylethyl isocyanate
Formule: C₉H₉NO
Kookpunt: 74 - 76 °C (7 hPa)
Dichtheid: 1,05 g/cm³ (20 °C)
Vlampunt: 65°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00063015
CAS nummer: 33375-06-3
EINECS: 251-485-2
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e89b893130dab9d5fee1e0204ad7c3bc4d6e881&vid=5e89b893130dab9d5fee1e0204ad7c3bc4d6e881&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR