Trichloroacetyl isocyanate ≥97.0% (by total nitrogen basis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
T1047-1G
T1047-25G
T1047-5G
TCIAT1047-1GEA
26.4
EUR
TCIAT1047-1G
TCIAT1047-25G
TCIAT1047-5G
Trichloroacetyl isocyanate ≥97.0% (by total nitrogen basis)
Trichloroacetyl isocyanate
Formule:
C₃Cl₃NO₂ Kookpunt: 80 - 85 °C (27 hPa) Dichtheid: 1,59 g/cm³ (20 °C) Vlampunt: 65°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00002033 CAS nummer: 3019-71-4 EINECS: 221-165-7 UN: 2206 ADR: 6.1,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...