Methyl decanoate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Decanoic acid methyl ester, Capric acid methyl ester, Decylic acid methyl ester, Methyl caprate
D0023-25ML D0023-500ML
TCIAD0023-25MLEA 17.6 EUR
TCIAD0023-25ML TCIAD0023-500ML
Methyl decanoate ≥98.0% (by GC)
Methyl decanoate
Formule: C₁₁H₂₂O₂
Kookpunt: 229°C
Smeltpunt: -18°C
Dichtheid: 0,8724 g/cm³ (20 °C)
Vlampunt: 107°C
MDL Nummer: MFCD00009580
CAS nummer: 110-42-9
EINECS: 203-766-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7424a47805fb40486ff9ce5aebac84e777fb4e06&vid=7424a47805fb40486ff9ce5aebac84e777fb4e06&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %