2-Ethylbutyl acetate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A0029-25ML
A0029-500ML
TCIAA0029-25MLEA
27.5
EUR
TCIAA0029-25ML
TCIAA0029-500ML
2-Ethylbutyl acetate ≥98.0% (by GC)
2-Ethylbutyl acetate
Formule:
C₈H₁₆O₂ Kookpunt: 159°C Dichtheid: 0,8784 g/cm³ (20 °C) Vlampunt: 53°C |
MDL Nummer:
MFCD00009343 CAS nummer: 10031-87-5 EINECS: 233-095-4 UN: 1177 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...