2-Ethylhexyl acetate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A0031-25ML
A0031-500ML
TCIAA0031-25MLEA
29.7
EUR
TCIAA0031-25ML
TCIAA0031-500ML
2-Ethylhexyl acetate ≥98.0% (by GC)
2-Ethylhexyl acetate
Formule:
C₁₀H₂₀O₂ Kookpunt: 200°C Smeltpunt: -80°C Dichtheid: 0,8726 g/cm³ (20 °C) Vlampunt: 77°C |
MDL Nummer:
MFCD00027249 CAS nummer: 103-09-3 EINECS: 203-079-1 Merck Index: 14,06763 |
|
Specification Test Results
Appearance | Colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...