7-Aminoenanthic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
7-Aminoheptanoic acid, 7-Aminoheptylic acid
A0311-1G
A0311-25G
A0311-5G
TCIAA0311-1GEA
47.3
EUR
TCIAA0311-1G
TCIAA0311-25G
TCIAA0311-5G
7-Aminoenanthic acid ≥98.0% (by GC, titration analysis)
7-Aminoenanthic acid
Formule:
C₇H₁₅NO₂ MW: 145.2 g/mol |
MDL Nummer:
MFCD00008242 CAS nummer: 929-17-9 EINECS: 213-197-5 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...