1,1,3,3-Tetramethoxypropane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Malonaldehyde bis(dimethyl acetal)

M0544-25G M0544-500G
TCIAM0544-25GEA 17.6 EUR
TCIAM0544-25G TCIAM0544-500G
1,1,3,3-Tetramethoxypropane ≥98.0% (by GC)
1,1,3,3-Tetramethoxypropane
Formule: C₇H₁₆O₄
MW: 164.2 g/mol
Kookpunt: 78°C
Vlampunt: 63°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008488
CAS nummer: 102-52-3
EINECS: 203-037-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15ab1290cfce2650e852c9e8db42c4fd55a17748&vid=15ab1290cfce2650e852c9e8db42c4fd55a17748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR