Itaconic acid ≥99.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M0223-25G M0223-500G
TCIAM0223-25GEA 18.7 EUR
TCIAM0223-25G TCIAM0223-500G
Itaconic acid ≥99.0% (by titrimetric analysis)
Itaconic acid
Formule: C₅H₆O₄
Kookpunt: 381 °C (1013 hPa)
Smeltpunt: 162…167 °C
Dichtheid: 1,573 g/cm³ (20 °C)
MDL Nummer: MFCD00004260
CAS nummer: 97-65-4
EINECS: 202-599-6
Merck Index: 14,05242
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=986274e9bae058818059db72907a2206e7162ac9&vid=986274e9bae058818059db72907a2206e7162ac9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 165.0 to 169.0 deg-C
Solubility in Water very faint turbidity