Sodium-2-ethylhexanoate ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2-Ethylcaproic acid sodium salt, Sodium 2-ethylhexanoate, 2-Ethylhexanoic acid sodium salt
S0862-25G
S0862-250G
TCIAS0862-25GEA
17.6
EUR
TCIAS0862-25G
TCIAS0862-250G
Sodium-2-ethylhexanoate ≥98.0% (by titrimetric analysis)
Sodium-2-ethylhexanoate
Formule:
C₈H₁₅NaO₂ MW: 166.2 g/mol |
MDL Nummer:
MFCD00014007 CAS nummer: 19766-89-3 EINECS: 243-283-8 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...