2-Bromo-6-chloro-4-fluoroaniline ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B3681-25G
B3681-5G
TCIAB3681-25GEA
250
EUR
TCIAB3681-25G
TCIAB3681-5G
2-Bromo-6-chloro-4-fluoroaniline ≥97.0% (by GC)
2-Bromo-6-chloro-4-fluoroaniline
Formule:
C₆H₄BrClFN Kookpunt: 260 °C (1013 hPa) Smeltpunt: 55…56 °C |
MDL Nummer:
MFCD00051707 CAS nummer: 201849-14-1 ADR: 6.1,III |
|
Specification Test Results
Appearance | White to Brown to Dark purple powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...