2-Chloor-6-fluorfenol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C2049-25G C2049-5G
TCIAC2049-25GEA 184 EUR
TCIAC2049-25G TCIAC2049-5G
2-Chloor-6-fluorfenol ≥98.0% (by GC)
2-Chloor-6-fluorfenol
Formule: C₆H₄ClFO
Kookpunt: 199 °C (1013 hPa)
Smeltpunt: 65 °C
MDL Nummer: MFCD01631574
CAS nummer: 2040-90-6
EINECS: 433-890-8
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1758af2a08e5907cd039b9ab3c5d3b40f5d88969&vid=1758af2a08e5907cd039b9ab3c5d3b40f5d88969&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR