5-Nitroisophthalic acid ≥99.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
N0520-25G
N0520-500G
TCIAN0520-25GEA
24.2
EUR
TCIAN0520-25G
TCIAN0520-500G
5-Nitroisophthalic acid ≥99.0% (by HPLC)
5-Nitroisophthalic acid
Formule:
C₈H₅NO₆ Smeltpunt: 260…264 °C |
MDL Nummer:
MFCD00007254 CAS nummer: 618-88-2 EINECS: 210-568-3 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...