2,3-Difluoro-6-nitrophenol ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1,2-Difluoro-3-hydroxy-4-nitrobenzene

D2705-25G D2705-5G
TCIAD2705-25GEA 213 EUR
TCIAD2705-25G TCIAD2705-5G
2,3-Difluoro-6-nitrophenol ≥98.0% (by GC, titration analysis)
2,3-Difluoro-6-nitrophenol
Formule: C₆H₃F₂NO₃
Smeltpunt: 61 °C
MDL Nummer: MFCD00042255
CAS nummer: 82419-26-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=182a8aff0d0abf52af1bfe3836b8e59742cb6284&vid=182a8aff0d0abf52af1bfe3836b8e59742cb6284&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 61.0 to 65.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR