Benzeneselenenyl chloride ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

P1091-25G P1091-5G
TCIAP1091-25GEA 215 EUR
TCIAP1091-25G TCIAP1091-5G
Benzeneselenenyl chloride ≥97.0% (by titrimetric analysis)
Benzeneselenenyl chloride
Formule: (C₆H₅)SeCl
Kookpunt: 95°C
Smeltpunt: 61…62 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000478
CAS nummer: 5707-04-0
EINECS: 227-196-2
UN: 2928
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdf1a3f37291c48ae3f981a9a805585e94e47e43&vid=cdf1a3f37291c48ae3f981a9a805585e94e47e43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Iodometric Titration) min. 97.0 %
Melting point 59.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR