ω-Bromo-2-nitroacetophenone ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B3528-25G
B3528-5G
TCIAB3528-25GEA
255
EUR
TCIAB3528-25G
TCIAB3528-5G
ω-Bromo-2-nitroacetophenone ≥98.0% (by GC)
ω-Bromo-2-nitroacetophenone
Formule:
C₈H₆BrNO₃ |
MDL Nummer:
MFCD00010294 CAS nummer: 6851-99-6 UN: 3261 ADR: 8,II |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...