2'-Fluoro-4'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
F0418-1G
F0418-5G
TCIAF0418-1GEA
22
EUR
TCIAF0418-1G
TCIAF0418-5G
2'-Fluoro-4'-hydroxyacetophenone ≥98.0% (by GC, titration analysis)
2'-Fluoro-4'-hydroxyacetophenone
Formule:
C₈H₇FO₂ |
MDL Nummer:
MFCD00142713 CAS nummer: 98619-07-9 |
|
Specification Test Results
Appearance | White to Light yellow to Light red powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...