4'-Phenylacetophenone ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A1025-25G A1025-250G
TCIAA1025-25GEA 22 EUR
TCIAA1025-25G TCIAA1025-250G
4'-Phenylacetophenone ≥98.0% (by GC)
4'-Phenylacetophenone
Formule: C₁₄H₁₂O
Kookpunt: 168°C
Smeltpunt: 118…121 °C
Dichtheid: 1,251 g/cm³ (0 °C)
MDL Nummer: MFCD00008749
CAS nummer: 92-91-1
EINECS: 202-202-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25722b7e00f16371c69e2b9c05e2cf110fedc583&vid=25722b7e00f16371c69e2b9c05e2cf110fedc583&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C