Cyclohexanone oxime ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C0490-25G C0490-500G
TCIAC0490-25GEA 18.7 EUR
TCIAC0490-25G TCIAC0490-500G
Cyclohexanone oxime ≥98.0% (by GC)
Cyclohexanone oxime
Formule: C₆H₁₁NO
Kookpunt: 210°C
Smeltpunt: 88…90 °C
Dichtheid: 1,13 g/cm³ (20 °C)
MDL Nummer: MFCD00001660
CAS nummer: 100-64-1
EINECS: 202-874-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97c40d9f54f3ee927f01c1c021cc863e9d8429af&vid=97c40d9f54f3ee927f01c1c021cc863e9d8429af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 92.0 deg-C
Solubility in Methanol almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR