Cyclopropyl methyl ketone ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C0985-25ML C0985-500ML C0985-100ML
TCIAC0985-25MLEA 31.9 EUR
TCIAC0985-25ML TCIAC0985-500ML TCIAC0985-100ML
Cyclopropyl methyl ketone ≥98.0% (by GC)
Cyclopropyl methyl ketone
Formule: C₅H₈O
Kookpunt: 110°C
Smeltpunt: –68 °C
Dichtheid: 0,899 g/cm³ (20 °C)
Vlampunt: 13°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001297
CAS nummer: 765-43-5
EINECS: 212-146-4
UN: 1224
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d83d04a3f4532b09e89240c07570a7ec9929d18f&vid=d83d04a3f4532b09e89240c07570a7ec9929d18f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %