Tetrabutylammonium hexafluorophosphate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Tetrabutylammonium hexafluorophosphate(V), Tetra-n-butylammonium hexafluorophosphate

T1279-25G T1279-250G
TCIAT1279-25GEA 30.8 EUR
TCIAT1279-25G TCIAT1279-250G
Tetrabutylammonium hexafluorophosphate ≥98.0% (by titrimetric analysis)
Tetrabutylammonium hexafluorophosphate
Formule: C₁₆H₃₆F₆NP
MW: 387.43 g/mol
Smeltpunt: 245°C
MDL Nummer: MFCD00011748
CAS nummer: 3109-63-5
EINECS: 221-472-6
UN: 1759
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8957bd1b668522b57fa4f5dee186d43005f301c&vid=c8957bd1b668522b57fa4f5dee186d43005f301c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR