6-Bromo-2-naphthol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
6-Bromo-2-hydroxynaphthalene, 2-Bromo-6-hydroxynaphthalene
B0621-25G
B0621-100G
TCIAB0621-25GEA
35.2
EUR
TCIAB0621-25G
TCIAB0621-100G
6-Bromo-2-naphthol ≥98.0% (by GC)
6-Bromo-2-naphthol
Formule:
C₁₀H₇BrO Kookpunt: 200 - 205 °C (20 -rr) Smeltpunt: 128 °C |
MDL Nummer:
MFCD00004081 CAS nummer: 15231-91-1 EINECS: 239-279-0 |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 127.0 to 134.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...