Maltol ≥99.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M0673-25G M0673-500G M0673-100G
TCIAM0673-25GEA 17.6 EUR
TCIAM0673-25G TCIAM0673-500G TCIAM0673-100G
Maltol ≥99.0% (by HPLC, titration analysis)
Maltol
Formule: C₆H₆O₃
Kookpunt: 285 °C (1013 hPa)
Dichtheid: 1,453 g/cm³ (20 °C)
MDL Nummer: MFCD00006578
CAS nummer: 118-71-8
EINECS: 204-271-8
Merck Index: 14,05713
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ae22f9fa0843021eb210c59ea88b6e15c4279c0&vid=3ae22f9fa0843021eb210c59ea88b6e15c4279c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 161.0 to 165.0 deg-C
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR