2-Acetyl-3-ethylpyrazine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A2003-1G A2003-5G
TCIAA2003-1GEA 17.6 EUR
TCIAA2003-1G TCIAA2003-5G
2-Acetyl-3-ethylpyrazine ≥98.0% (by GC)
2-Acetyl-3-ethylpyrazine
Formule: C₈H₁₀N₂O
Kookpunt: 77°C
Vlampunt: 87°C
MDL Nummer: MFCD00038028
CAS nummer: 32974-92-8
EINECS: 251-316-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a373edb747728fadc040924507501ab0aebaa247&vid=a373edb747728fadc040924507501ab0aebaa247&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %