2-Bromo-4-nitropyridine ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3523-1G B3523-5G
TCIAB3523-1GEA 59.4 EUR
TCIAB3523-1G TCIAB3523-5G
2-Bromo-4-nitropyridine ≥95.0% (by GC)
2-Bromo-4-nitropyridine
Formule: C₅H₃BrN₂O₂
MW: 203 g/mol
MDL Nummer: MFCD03701383
CAS nummer: 6945-67-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae6da7af643f6f9193e28ba7060794f3e7612bcc&vid=ae6da7af643f6f9193e28ba7060794f3e7612bcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR