2,4,6-Triphenyl-1,3,5-triazine ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
T2785-1G
T2785-5G
TCIAT2785-1GEA
18.7
EUR
TCIAT2785-1G
TCIAT2785-5G
2,4,6-Triphenyl-1,3,5-triazine ≥98.0% (by GC)
2,4,6-Triphenyl-1,3,5-triazine
Formule:
C₂₁H₁₅N₃ Kookpunt: 230°C Smeltpunt: 243 °C Dichtheid: 1,335 g/cm³ (20 °C) |
MDL Nummer:
MFCD00006051 CAS nummer: 493-77-6 EINECS: 207-779-8 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 234.0 to 238.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...