Lecithin from egg yolk
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L0022-25G L0022-100G
TCIAL0022-25GEA 46.2 EUR
TCIAL0022-25G TCIAL0022-100G
Lecithin from egg yolk
Lecithin from egg yolk
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00131449
CAS nummer: 8002-43-5
EINECS: 232-307-2
Merck Index: 14,05429
Order Now
Specification Test Results
Appearance Orange to Brown clear liquid to cloudy liquid
Saponification Value 180.0 to 200.0
Acid Value max. 10.0
Water max. 2.0 %