Methyl glycolate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Hydroxyacetic Acid Methyl Ester, Methyl 2-hydroxyacetate, Glycolic Acid Methyl Ester, Methyl Hydroxyacetate
G0199-25G
G0199-500G
G0199-100G
TCIAG0199-25GEA
53.9
EUR
TCIAG0199-25G
TCIAG0199-500G
TCIAG0199-100G
Methyl glycolate ≥98.0% (by GC)
Methyl glycolate
Formule:
C₃H₆O₃ Kookpunt: 151°C Smeltpunt: 54…56 °C Dichtheid: 1,1756 g/cm³ (20 °C) Vlampunt: 67°C |
MDL Nummer:
MFCD00004667 CAS nummer: 96-35-5 EINECS: 202-502-7 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...