meso-2,3-Dimercaptosuccinic acid ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (R,S)-2,3-Dimercaptobutanedioic acid, meso-2,3-Dimercaptobutanedioic acid, Succimer, (R,S)-2,3-Dimercaptosuccinic acid, (2R,3S)-rel-2,3-Dimercaptosuccinic acid

D1722-25G D1722-5G
TCIAD1722-25GEA 293 EUR
TCIAD1722-25G TCIAD1722-5G
meso-2,3-Dimercaptosuccinic acid ≥95.0% (by titrimetric analysis)
meso-2,3-Dimercaptosuccinic acid
Formule: C₄H₆O₄S₂
MW: 182.22 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00064799
CAS nummer: 304-55-2
EINECS: 206-155-2
Merck Index: 12,09034
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fa3535e105e5fb9dbb2d3d4e97441cec9a5a2c9&vid=2fa3535e105e5fb9dbb2d3d4e97441cec9a5a2c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR